Learn More
Description
Specifications
Specifications
| Antigen | ATF2 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/ml, Flow Cytometry |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | NP_001871 |
| Gene Alias | activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558 |
| Gene Symbols | ATF2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
