Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF2, p Ser112, p Ser94 Antibody, Novus Biologicals™
SDP

Catalog No. NBP254642 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254642 100 μL
NBP254641 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP254642 Supplier Novus Biologicals Supplier No. NBP254642

Rabbit Polyclonal Antibody

ATF2 p Ser112, p Ser94 Polyclonal antibody specifically detects ATF2 p Ser112, p Ser94 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATF2 (p Ser112, p Ser94)
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:50-1:100
Formulation Saline (without Mg2+ abd Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol with 0.02% Sodium Azide
Gene Alias activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558
Gene Symbols ATF2
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of serine 112 or 94 (D-L-S(p)-P-L) derived from HumanATF2.
Molecular Weight of Antigen 70 kDa
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1386
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.