Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF2, p Thr71 Antibody (103C411.2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N233236J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N233236J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N233236J646 Supplier Novus Biologicals Supplier No. NBP233236JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATF2 Monoclonal specifically detects ATF2 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen ATF2
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 103C411.2
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558
Gene Symbols ATF2
Host Species Mouse
Immunogen This antibody was generated by immunizing mice with a synthetic peptide containing phosphorylated threonine at position 71 of human ATF-2.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Chromatin Research, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1386.0
Test Specificity This monoclonal antibody is specific for phospho (Thr71) ATF2. It cannot be used to test for non-phosphorylated ATF2.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.