Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP102935SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP102935SS 0.025 mL
NBP102935 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP102935SS Supplier Novus Biologicals Supplier No. NBP102935SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

ATF3 Polyclonal specifically detects ATF3 in Human, Mouse, Rat, Mammal samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATF3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. P18847
Gene Alias activating transcription factor 3cAMP-dependent transcription factor ATF-3, ATF3deltaZip2, ATF3deltaZip2c, ATF3deltaZip3, cyclic AMP-dependent transcription factor ATF-3
Gene Symbols ATF3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a region of human ATF3 (within residues 100-150). [UniProt# P18847]
Molecular Weight of Antigen 17 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 467
Test Specificity Human, mouse and rat. Predicted to react with most mammalian species based on sequence homology.
Target Species Human, Mouse, Rat, Mammalia
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.