Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF5 Rabbit anti-Human, Mouse, Rat, Clone: 9Z3M0, Novus Biologicals™
SDP

Catalog No. NB167534 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167534 20 μg
NB167533 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167534 Supplier Novus Biologicals Supplier No. NBP31622120UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ATF5 Monoclonal antibody specifically detects ATF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ATF5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9Z3M0
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias activating transcription factor 5FLJ34666, ATFXHMFN0395, cAMP-dependent transcription factor ATF-5, cyclic AMP-dependent transcription factor ATF-5, Transcription factor ATFx
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 183-282 of human ATF5 (Q9Y2D1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 22809
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.