Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF6 Antibody (70B1413.1), PE, Novus Biologicals™
SDP

Catalog No. NBP140256PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP140256PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP140256PE Supplier Novus Biologicals Supplier No. NBP140256PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATF6 Monoclonal specifically detects ATF6 in Human, Mouse, Rat, Porcine, Rabbit samples. It is validated for Flow Cytometry, Flow (Intracellular), Knockout Validated.

Specifications

Antigen ATF6
Applications Flow Cytometry, Immunoassay
Classification Monoclonal
Clone 70B1413.1
Conjugate PE
Dilution Flow Cytometry 1 μg/million cells, Flow (Intracellular), Knockout Validated
Gene Accession No. P18850
Gene Alias activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha
Gene Symbols ATF6
Host Species Mouse
Immunogen This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 22926
Test Specificity This ATF6 antibody detects both the full length and the cleaved/active protein.
Target Species Human, Mouse, Rat, Pig, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.