Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF6 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP175478SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175478SS 0.025 mL
NBP175478 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP175478SS Supplier Novus Biologicals Supplier No. NBP175478SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

ATF6 Polyclonal specifically detects ATF6 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ATF6
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Simple Western 1:25, Immunocytochemistry/Immunofluorescence 1:100
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. P18850
Gene Alias activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha
Gene Symbols ATF6
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human ATF6 protein (within residues 200-350). [Swiss-Prot P18850]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 22926
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.