Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF7 Antibody, Novus Biologicals™
SDP

Catalog No. NB300899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300899 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300899 Supplier Novus Biologicals Supplier No. NB300899
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ATF7 Polyclonal specifically detects ATF7 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen ATF7
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006847.
Gene Alias activating transcription factor 7ATFATranscription factor ATF-A, cAMP-dependent transcription factor ATF-7, cyclic AMP-dependent transcription factor ATF-7, MGC57182
Gene Symbols ATF7
Host Species Goat
Immunogen Peptide with sequence C-HVIMTPQSQSAGR corresponding to C-Terminus according to NP_001123531.1, NP_006847.1, NP_001123532.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11016
Test Specificity This antibody is expected to recognize all three reported isoforms (NP_001123531.1, NP_006847.1 and NP_001123532.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.