Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG101 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB163696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163696 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163696 Supplier Novus Biologicals Supplier No. NBP281705AF647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATG101 Polyclonal antibody specifically detects ATG101 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ATG101
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias ATG101, Atg13-interacting protein, autophagy-related protein 101, C12orf44, chromosome 12 open reading frame 44, FLJ11773
Host Species Rabbit
Immunogen ATG101 antibody was raised against a 16 amino acid synthetic peptide near the center of human ATG101. The immunogen is located within amino acids 80 - 130 of ATG101. Amino Acid Squence: CSLEFYQKKKSRWPFSD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 60673
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.