Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG11 Antibody (7C2NB), Novus Biologicals™
SDP

Catalog No. NBP247124SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247124SS 0.025 mg
NBP247124 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP247124SS Supplier Novus Biologicals Supplier No. NBP247124SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATG11 Monoclonal specifically detects ATG11 in Yeast samples. It is validated for Western Blot.

Specifications

Antigen ATG11
Applications Western Blot
Classification Monoclonal
Clone 7C2NB
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias CVT3, CVT9
Gene Symbols ATG11
Host Species Mouse
Immunogen Partial recombinant protein made to an internal sequence of the S. cerevisiae ATG11 protein (between amino acids 50-250) [UniProt Q12527]
Purification Method IgM purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 856162
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.