Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG11 Antibody (7C2NB), Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $402.00
Specifications
Antigen | ATG11 |
---|---|
Clone | 7C2NB |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5 ug/ml |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247124SS
![]() |
Novus Biologicals
NBP247124SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP247124
![]() |
Novus Biologicals
NBP247124 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
ATG11 Monoclonal specifically detects ATG11 in Yeast samples. It is validated for Western Blot.Specifications
ATG11 | |
1.0 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Autophagy | |
PBS with 0.05% Sodium Azide | |
856162 | |
Partial recombinant protein made to an internal sequence of the S. cerevisiae ATG11 protein (between amino acids 50-250) [UniProt Q12527] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
7C2NB | |
Western Blot 0.5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Yeast | |
CVT3, CVT9 | |
ATG11 | |
IgM λ | |
IgM purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title