Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG12 Antibody (2F6), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32649550UL
Description
ATG12 Monoclonal antibody specifically detects ATG12 in Human samples. It is validated for ELISASpecifications
ATG12 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
2F6 | |
ELISA | |
Apg12 (autophagy 12, S. cerevisiae)-like, APG12 autophagy 12-like (S. cerevisiae), APG12HAPG12, APG12-like, ATG12 autophagy related 12 homolog (S. cerevisiae), Autophagy-related protein 12, FBR93, ubiquitin-like protein ATG12, yeast) homolog | |
A synthesized peptide derived from Human ATG12 [UniProt O94817] | |
50 μL | |
Apoptosis, Autophagy, Neuroscience | |
9140 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction