Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATG12 Antibody (2F6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | ATG12 |
---|---|
Clone | 2F6 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
ATG12 Monoclonal antibody specifically detects ATG12 in Human samples. It is validated for ELISASpecifications
ATG12 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Apg12 (autophagy 12, S. cerevisiae)-like, APG12 autophagy 12-like (S. cerevisiae), APG12HAPG12, APG12-like, ATG12 autophagy related 12 homolog (S. cerevisiae), Autophagy-related protein 12, FBR93, ubiquitin-like protein ATG12, yeast) homolog | |
A synthesized peptide derived from Human ATG12 [UniProt O94817] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2F6 | |
ELISA | |
Unconjugated | |
Rabbit | |
Apoptosis, Autophagy, Neuroscience | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
9140 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title