Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG12 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB195397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB195397 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB195397 Supplier Novus Biologicals Supplier No. NBP176861DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG12 Polyclonal antibody specifically detects ATG12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ATG12
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Apg12 (autophagy 12, S. cerevisiae)-like, APG12 autophagy 12-like (S. cerevisiae), APG12HAPG12, APG12-like, ATG12 autophagy related 12 homolog (S. cerevisiae), Autophagy-related protein 12, FBR93, ubiquitin-like protein ATG12, yeast) homolog
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the center of human ATG12. The immunogen is located within the first 50 amino acids of ATG12. Amino Acid Squence: LTDVSPETTTPEPPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9140
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.