Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG13, p Ser355 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219127SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219127SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219127SS Supplier Novus Biologicals Supplier No. NBP219127SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 18 publications

ATG13 Polyclonal specifically detects ATG13 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Knockdown Validated.

Specifications

Antigen ATG13 (p Ser355)
Applications Western Blot, Flow Cytometry, ELISA
Classification Polyclonal
Concentration 1.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:25000-1:175000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. O75143
Gene Alias autophagy-related protein 13, autophagy related 13, KIAA0652
Gene Symbols ATG13
Host Species Rabbit
Immunogen ATG13 [p Ser355] Antibody was prepared by repeated immunizations with a synthetic peptide corresponding to the region near Ser355 of ATG13. (Uniprot: O75143)
Molecular Weight of Antigen 56.57 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 9776
Test Specificity This affinity-purified antibody is directed against the phosphorylated form of human ATG13 protein at the pS318 residue. The resultant affinity purified antibody was then cross adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against human ATG13 pS318 protein and the antibody is specific for the phosphorylated form of the protein. Reactivity with non-phosphorylated human ATG13 is minimal by ELISA and western blot. A BLAST analysis was used to suggest cross reactivity with ATG13 from human based on 100% sequence homology with the immunogen. Reactivity against homologues from other sources is not known.
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.