Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG14 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB355551 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355551 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355551 Supplier Novus Biologicals Supplier No. NBP236445MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG14 Polyclonal antibody specifically detects ATG14 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ATG14
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ATG14 Autophagy Related 14 Homolog, ATG14 Autophagy Related 14 Homolog (S. Cerevisiae), ATG14L, Autophagy Related 14, Autophagy-Related Protein 14-Like Protein, Barkor, Beclin 1-Associated Autophagy-Related Key Regulator, Beclin 1-interacting protein, KIAA0831
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human ATG14. The immunogen is located within amino acids 270 - 320 of ATG14. Amino Acid Squence: VEEKKTTQGPDMEQSNPA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22863
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.