Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093361 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093361 20 μL
NB093360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093361 Supplier Novus Biologicals Supplier No. NBP30466420UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG14 Polyclonal specifically detects ATG14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATG14
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ATG14 Autophagy Related 14 Homolog, ATG14 Autophagy Related 14 Homolog (S. Cerevisiae), ATG14L, Autophagy Related 14, Autophagy-Related Protein 14-Like Protein, Barkor, Beclin 1-Associated Autophagy-Related Key Regulator, Beclin 1-interacting protein, KIAA0831
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human ATG14 (NP_055739.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22863
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.