Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG16L1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB11060928B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060928B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11060928B Supplier Novus Biologicals Supplier No. NB11060928B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG16L1 Polyclonal specifically detects ATG16L1 in Human, Mouse, Rat, Alligator, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Electron Microscopy.

Specifications

Antigen ATG16L1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Electron Microscopy
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q676U5
Gene Alias APG16 autophagy 16-like (S. cerevisiae), APG16L beta, APG16LFLJ10828, APG16-like 1, ATG16 autophagy related 16-like (S. cerevisiae), ATG16 autophagy related 16-like 1 (S. cerevisiae), ATG16A, ATG16L, autophagy-related protein 16-1, FLJ00045, FLJ10035, FLJ22677, IBD10, WD repeat domain 30, WDR30
Gene Symbols ATG16L1
Host Species Rabbit
Immunogen A synthetic peptide within residues 1-100 of human ATG16L1 protein. [Swiss-Prot# Q676U5]
Molecular Weight of Antigen 68 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 55054
Target Species Human, Mouse, Rat, Alligator, Bovine, Canine, Primate
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.