Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG16L1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11082384T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11082384T 0.025 mL
NB11082384 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11082384T Supplier Novus Biologicals Supplier No. NB11082384SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

ATG16L1 Polyclonal specifically detects ATG16L1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ATG16L1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunoprecipitation reported in scientific literature (PMID 29361549)
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Alias APG16 autophagy 16-like (S. cerevisiae), APG16L beta, APG16LFLJ10828, APG16-like 1, ATG16 autophagy related 16-like (S. cerevisiae), ATG16 autophagy related 16-like 1 (S. cerevisiae), ATG16A, ATG16L, autophagy-related protein 16-1, FLJ00045, FLJ10035, FLJ22677, IBD10, WD repeat domain 30, WDR30
Gene Symbols ATG16L1
Host Species Rabbit
Immunogen A synthetic peptide within residues 1-100 of mouse ATG16L1. [Swiss-Prot# Q8C0J2]
Molecular Weight of Antigen 66 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 55054
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.