Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG16L1 Rabbit anti-Human, Mouse, Rat, Clone: JU30-62, Novus Biologicals™
SDP

Catalog No. NBP275413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275413 Supplier Novus Biologicals Supplier No. NBP275413

Rabbit Monoclonal Antibody

ATG16L1 Monoclonal specifically detects ATG16L1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ATG16L1
Applications Western Blot
Classification Monoclonal
Clone JU30-62
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-2000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias APG16 autophagy 16-like (S. cerevisiae), APG16L beta, APG16LFLJ10828, APG16-like 1, ATG16 autophagy related 16-like (S. cerevisiae), ATG16 autophagy related 16-like 1 (S. cerevisiae), ATG16A, ATG16L, autophagy-related protein 16-1, FLJ00045, FLJ10035, FLJ22677, IBD10, WD repeat domain 30, WDR30
Gene Symbols ATG16L1
Host Species Rabbit
Immunogen Recombinant protein within Nterminal Human ATG16L1. (SwissProt: Q676U5 Human; SwissProt: Q8C0J2 Mouse)
Molecular Weight of Antigen 68 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Autophagy, Cancer, Cell Biology, Macroautophagy, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 55054.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.