Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG2B Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB163712 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163712 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163712 Supplier Novus Biologicals Supplier No. NBP281706AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATG2B Polyclonal antibody specifically detects ATG2B in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen ATG2B
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias ATG2 autophagy related 2 homolog B (S. cerevisiae), C14orf103, FLJ10242
Host Species Rabbit
Immunogen ATG2B antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human ATG2B. The immunogen is located within amino acids 450 - 500 of ATG2B. Amino Acid Squence: EFLDHHKEQPVRGSTFPSN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Macroautophagy
Primary or Secondary Primary
Gene ID (Entrez) 55102
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.