Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG3/APG3 Antibody (OTI3H2), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N272140J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272140J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272140J549 Supplier Novus Biologicals Supplier No. NBP272140JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATG3/APG3 Monoclonal specifically detects ATG3/APG3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ATG3/APG3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3H2
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias APG3, APG3 autophagy 3-like (S. cerevisiae), APG3L, APG3-LIKE, ATG3 autophagy related 3 homolog (S. cerevisiae), Autophagy-related protein 3, DKFZp564M1178, EC 6.3.2.-, FLJ22125,2610016C12Rik, hApg3, MGC15201, PC3-96, Protein PC3-96, ubiquitin-like-conjugating enzyme ATG3
Gene Symbols ATG3
Host Species Mouse
Immunogen Full length recombinant protein of human ATG3/APG3 (NP_071933) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Autophagy, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 64422.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.