Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG4A Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB342395 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342395 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342395 Supplier Novus Biologicals Supplier No. NBP281788MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG4A Polyclonal antibody specifically detects ATG4A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ATG4A
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias APG4 autophagy 4 homolog A (S. cerevisiae), APG4A, ATG4 autophagy related 4 homolog A (S. cerevisiae), AUTL2, AUT-like 2 cysteine endopeptidase, AUT-like 2, cysteine endopeptidase (S. cerevisiae), autophagin 2, autophagin-2, cysteine endopeptidase, cysteine protease ATG4A, EC 3.4.22, EC 3.4.22.-, hAPG4A
Host Species Rabbit
Immunogen ATG4A antibody was raised against an 18 amino acid peptide near the carboxy terminus of human ATG4A. The immunogen is located within the last 50 amino acids of ATG4A. Amino Acid Squence: VQKHPSHWPPFVPPAKPE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 115201
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.