Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG4B Antibody, Novus Biologicals™
SDP

Catalog No. NBP224709SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224709SS 0.025 mg
NBP224709 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224709SS Supplier Novus Biologicals Supplier No. NBP224709SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG4B Polyclonal specifically detects ATG4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATG4B
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias APG4 autophagy 4 homolog B, APG4 autophagy 4 homolog B (S. cerevisiae), APG4B, ATG4 autophagy related 4 homolog B (S. cerevisiae), AUTL1MGC1353, AUT-like 1 cysteine endopeptidase, Autophagin-1, Autophagy-related cysteine endopeptidase 1, Autophagy-related protein 4 homolog B, cysteine protease ATG4B, DKFZp586D1822, EC 3.4.22, EC 3.4.22.-, hAPG4B, KIAA0943
Gene Symbols ATG4B
Host Species Rabbit
Immunogen A portion of amino acid 350-400 of human APG4B was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Autophagy, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 23192
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, monkey, rat, dog and orangutan, 94% homologous in opossum and mouse, 88% in zebrafish, pig, and catfish.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.