Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG4C Rabbit anti-Human, Clone: S08-8G9, Novus Biologicals™
SDP

Catalog No. NB188361 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188361 100 μg
NB188362 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188361 Supplier Novus Biologicals Supplier No. NBP315003100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ATG4C Monoclonal antibody specifically detects ATG4C in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence

Specifications

Antigen ATG4C
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S08-8G9
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin, Immunofluorescence 20 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias APG4 autophagy 4 homolog C, APG4 autophagy 4 homolog C (S. cerevisiae), APG4C, ATG4 autophagy related 4 homolog C (S. cerevisiae), AUT (S. cerevisiae)-like 1, cysteine endopeptidase; AUT-like 1, cysteineendopeptidase (S. cerevisiae), AUTL1, AUTL3APG4-C, AUT-like 1, cysteine endopeptidase, AUT-like 3 cysteine endopeptidase, Autophagin-3, Autophagy-related cysteine endopeptidase 3, Autophagy-related protein 4 homolog C, cysteine protease ATG4C, EC 3.4.22, EC 3.4.22.-, FLJ14867
Host Species Rabbit
Immunogen Recombinant protein of human ATG4C (Uniprot#: Q96DT6)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 84938
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.