Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG4C Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095974 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095974 100 μL
NB095975 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095974 Supplier Novus Biologicals Supplier No. NBP304503100UL

Rabbit Polyclonal Antibody

ATG4C Polyclonal specifically detects ATG4C in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ATG4C
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias APG4 autophagy 4 homolog C, APG4 autophagy 4 homolog C (S. cerevisiae), APG4C, ATG4 autophagy related 4 homolog C (S. cerevisiae), AUT (S. cerevisiae)-like 1, cysteine endopeptidase; AUT-like 1, cysteineendopeptidase (S. cerevisiae), AUTL1, AUTL3APG4-C, AUT-like 1, cysteine endopeptidase, AUT-like 3 cysteine endopeptidase, Autophagin-3, Autophagy-related cysteine endopeptidase 3, Autophagy-related protein 4 homolog C, cysteine protease ATG4C, EC 3.4.22, EC 3.4.22.-, FLJ14867
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ATG4C (NP_116241.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Autophagy, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 84938
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.