Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG5 Antibody (603813), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB296109 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296109 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296109 Supplier Novus Biologicals Supplier No. FAB5294MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATG5 Monoclonal specifically detects ATG5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.

Specifications

Antigen ATG5
Applications Immunohistochemistry, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 603813
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25
Host Species Mouse
Immunogen E. coli-derived recombinant human ATG5, Asn99-Thr193, Accession # Q9H1Y0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 9474
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.