Learn More
Description
Specifications
Specifications
| Antigen | ATG5 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, Radioimmune Assays (RIA), In situ PLA, Immunoassay, KnockDown |
| Classification | Polyclonal |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Simple Western 1:50, Flow Cytometry, ELISA, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:400, Immunoblotting, Radioimmunoassay, Proximity Ligation Assay, Electron Microscopy, Knockout Validated, KnockDown Validated |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q9H1Y0 |
| Gene Alias | APG5 autophagy 5-like (S. cerevisiae), ASPAPG5-LIKE, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5 |
| Gene Symbols | ATG5 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
