Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG5 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB186003 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186003 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186003 Supplier Novus Biologicals Supplier No. NBP311775F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATG5 Polyclonal antibody specifically detects ATG5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)

Specifications

Antigen ATG5
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Polyclonal
Conjugate FITC
Formulation PBS with 0.05% Sodium Azide
Gene Alias APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25
Host Species Rabbit
Immunogen A partial recombinant protein produced in E. coli made to an internal portion of the human ATG5 protein produce in (between amino acids 80-400) [UniProt Q9H1Y0].
Purification Method Immunogen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 9474
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.