Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG5 Antibody (rATG5/2553), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB386876 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386876 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386876 Supplier Novus Biologicals Supplier No. NBP308452G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATG5 Monoclonal antibody specifically detects ATG5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen ATG5
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone rATG5/2553
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias APG5, APG5 (Autophagy 5, S. Cerevisiae)-Like, APG5 autophagy 5-like (S. cerevisiae), APG5L, APG5-LIKE, ASP, ASPAPG5-LIKE, ATG5 Autophagy Related 5 Homolog, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5, Autophagy Related 5, HAPG5, SCAR25
Host Species Mouse
Immunogen Recombinant fragment of human ATG5 protein (around aa 1-119) (exact sequence is proprietary) (Uniprot: Q9H1Y0)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 9474
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.