Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG7 Antibody (683906) [PE], Novus Biologicals™
SDP

Catalog No. NB376136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376136 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376136 Supplier Novus Biologicals Supplier No. FAB6608P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATG7 Monoclonal antibody specifically detects ATG7 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen ATG7
Applications Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 683906
Conjugate PE
Dilution Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias APG7 autophagy 7-like (S. cerevisiae), APG7L, APG7-LIKE, ATG12-activating enzyme E1 ATG7, ATG7 autophagy related 7 homolog (S. cerevisiae), Autophagy-related protein 7, DKFZp434N0735, GSA7, hAGP7, ubiquitin activating enzyme E1-like protein, Ubiquitin-activating enzyme E1-like protein, ubiquitin-like modifier-activating enzyme ATG7
Host Species Mouse
Immunogen E. coli-derived recombinant human ATG7, Asn387-Gln570, Accession # O95352
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 10533
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.