Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG7 Rabbit anti-Human, Mouse, Clone: SC06-30, Novus Biologicals™
SDP

Catalog No. NBP267596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267596 Supplier Novus Biologicals Supplier No. NBP267596

Rabbit Monoclonal Antibody

ATG7 Monoclonal specifically detects ATG7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen ATG7
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SC06-30
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias APG7 autophagy 7-like (S. cerevisiae), APG7L, APG7-LIKE, ATG12-activating enzyme E1 ATG7, ATG7 autophagy related 7 homolog (S. cerevisiae), Autophagy-related protein 7, DKFZp434N0735, GSA7, hAGP7, ubiquitin activating enzyme E1-like protein, Ubiquitin-activating enzyme E1-like protein, ubiquitin-like modifier-activating enzyme ATG7
Gene Symbols ATG7
Host Species Rabbit
Immunogen Synthetic peptide within Human ATG7 aa 654-703/ 703. (SwissProt: O95352 Human; SwissProt: Q9D906 Mouse)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 10533
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.