Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG9A Antibody (Atg9 14F2 8B1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP250109AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250109AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250109AH Supplier Novus Biologicals Supplier No. NBP250109AF647

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Hamster Monoclonal Antibody

ATG9A Monoclonal specifically detects ATG9A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen ATG9A
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Clone Atg9 14F2 8B1
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208
Gene Symbols ATG9A
Host Species Hamster
Immunogen Conserved C-terminal synthetic peptide of Human ATG9A sequence
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 79065
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.