Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG9A Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB293551 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293551 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293551 Supplier Novus Biologicals Supplier No. NBP326074IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG9A Polyclonal antibody specifically detects ATG9A in Human samples. It is validated for Immunoprecipitation

Specifications

Antigen ATG9A
Applications Immunoprecipitation
Classification Polyclonal
Conjugate DyLight 755
Dilution Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias APG9 autophagy 9-like 1, APG9 autophagy 9-like 1 (S. cerevisiae), APG9L1APG9-like 1, ATG9 autophagy related 9 homolog A (S. cerevisiae), autophagy 9-like 1 protein, autophagy-related protein 9A, FLJ22169, mATG9, MGD3208
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ATG9A fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 79065
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.