Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG9B Antibody, Novus Biologicals™
SDP

Catalog No. NB017936 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017936 0.05 mL
NB10098679 0.15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017936 Supplier Novus Biologicals Supplier No. NB100986790.05ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG9B Polyclonal specifically detects ATG9B in Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen ATG9B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, Immunohistochemistry 1:200-1:2000, Immunohistochemistry-Paraffin 1:200-1:2000, Immunohistochemistry-Frozen 1:200-1:2000
Formulation Whole antisera with No Preservative
Gene Alias APG9L2APG9-like 2, ATG9 autophagy related 9 homolog B (S. cerevisiae), autophagy-related protein 9B, endothelial nitric oxide synthase antisense, nitric oxide synthase 3 antisense, Nitric oxide synthase 3-overlapping antisense gene protein, NOS3ASautophagy 9-like 2 protein, Protein sONE, SONEFLJ14885
Gene Symbols ATG9B
Host Species Rabbit
Immunogen A synthetic peptide from an internal part of drosophila autophagy-related protein 9B conjugated to an immunogenic carrier protein has been used as the immunogen.
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 285973
Test Specificity ATG9B.
Reconstitution Reconstitute with 0.15 ml sterilized water.
Target Species Drosophila
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.