Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG9B Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB356391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356391 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356391 Supplier Novus Biologicals Supplier No. NBP177169PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG9B Polyclonal antibody specifically detects ATG9B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ATG9B
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias APG9L2APG9-like 2, ATG9 autophagy related 9 homolog B (S. cerevisiae), autophagy-related protein 9B, endothelial nitric oxide synthase antisense, nitric oxide synthase 3 antisense, Nitric oxide synthase 3-overlapping antisense gene protein, NOS3ASautophagy 9-like 2 protein, Protein sONE, SONEFLJ14885
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ATG9B. The immunogen is located within amino acids 850 - 900 of ATG9B. Amino Acid Squence: CPSQPPSPDEEKPSWSSD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 285973
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.