Learn More
Description
Specifications
Specifications
| Antigen | ATIC |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | OTI1D2 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-2000, Flow Cytometry 1:100, CyTOF-ready |
| Formulation | Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative |
| Gene Alias | 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMPcyclohydrolase, 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotidetransformylase/inosinicase, AICAR, AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme, AICARFT, AICARFT/IMPCHASE, bifunctional purine biosynthesis protein PURH, IMPCHASE, phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase, PURHFLJ93545 |
| Gene Symbols | ATIC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
