Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATIC Antibody (OTI1D2), Biotin, Novus Biologicals™
SDP

Catalog No. NBP270220B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270220B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270220B Supplier Novus Biologicals Supplier No. NBP270220B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATIC Monoclonal specifically detects ATIC in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen ATIC
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1D2
Conjugate Biotin
Dilution Western Blot, Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMPcyclohydrolase, 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotidetransformylase/inosinicase, AICAR, AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme, AICARFT, AICARFT/IMPCHASE, bifunctional purine biosynthesis protein PURH, IMPCHASE, phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase, PURHFLJ93545
Gene Symbols ATIC
Host Species Mouse
Immunogen Full length recombinant protein of human ATIC (NP_004035) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 471.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.