Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atlastin-2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290896 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290896 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290896 Supplier Novus Biologicals Supplier No. NBP178733PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Atlastin-2 Polyclonal antibody specifically detects Atlastin-2 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen Atlastin-2
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias ADP-ribosylation factor-like 6 interacting protein 2, ADP-ribosylation factor-like protein 6-interacting protein 2, ADP-ribosylation-like factor 6 interacting protein 2, aip-2, ARL3IP2, ARL-6-interacting protein 2, ARL6IP2, atlastin GTPase 2, atlastin2, atlastin-2, EC 3.6.5.-, FLJ23293
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 533 and 583 of human Atlastin GTPase 2 using the numbering given in entry NP_001129145.1 (GeneID 64225).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64225
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.