Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATM Antibody, Novus Biologicals™
SDP

Catalog No. NB100104 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100104 0.05 mL
NB100104SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100104 Supplier Novus Biologicals Supplier No. NB100104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 92 publications

ATM Polyclonal specifically detects ATM in Canine, Human, Kangaroo, Mouse, Rat samples. It is validated for ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen ATM
Applications ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 28512243)
Formulation PBS, 50% Glycerol with 0.05% Sodium Azide
Gene Accession No. Q13315
Gene Alias AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated (includes complementation groups A, C and D), ataxia telangiectasia mutatedATD, ATC, ATDC, ATE, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1
Gene Symbols ATM
Host Species Rabbit
Immunogen A fragment of the human ATM protein corresponding to the C-terminus (within the last third of the protein sequence). [Uniprot: Q13315]
Molecular Weight of Antigen 350 kDa
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Hypoxia, p53 Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 472
Target Species Canine, Mouse, Human, Kangaroo, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.