Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATM Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 92 publications
$280.00 - $501.50
Specifications
Antigen | ATM |
---|---|
Concentration | 2.5 mg/mL |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 28512243) |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100104SS
![]() |
Novus Biologicals
NB100104SS |
0.025 mL |
Each for $280.00
|
|
|||||
NB100104
![]() |
Novus Biologicals
NB100104 |
0.05 mL |
Each for $501.50
|
|
|||||
Description
ATM Polyclonal specifically detects ATM in Human, Mouse, Rat, Canine, Kangaroo samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
ATM | |
Western Blot 1:500-1:1000, Flow Cytometry, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 28512243) | |
Polyclonal | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Hypoxia, p53 Pathway, Tumor Suppressors | |
PBS, 50% Glycerol with 0.05% Sodium Azide | |
AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated (includes complementation groups A, C and D), ataxia telangiectasia mutatedATD, ATC, ATDC, ATE, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1 | |
ATM | |
IgG | |
Affinity Purified | |
350 kDa |
2.5 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Canine, Kangaroo | |
Q13315 | |
472 | |
A fragment of the human ATM protein corresponding to the C-terminus (within the last third of the protein sequence). [Uniprot: Q13315] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title