Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATM, p Ser1981 Antibody (10H11.E12), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB100306W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100306W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100306W Supplier Novus Biologicals Supplier No. NB100306AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATM (p Ser1981) Monoclonal specifically detects ATM (p Ser1981) in Canine (Negative), Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen ATM (p Ser1981)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10H11.E12
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. Q13315
Gene Alias AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated (includes complementation groups A, C and D), ataxia telangiectasia mutatedATD, ATC, ATDC, ATE, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1
Gene Symbols ATM
Host Species Mouse
Immunogen Synthetic peptide made to a region surrounding the phosphorylated Serine 1981 of human ATM. [UniProt# Q13315]
Molecular Weight of Antigen 370 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Hypoxia, Modulation of DNA Pools, p53 Pathway, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 472
Test Specificity Human, mouse and rat. Does not work in canine per customer review.
Target Species Canine (Negative), Human, Rat, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.