Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATM, p Ser1981 Antibody (10H11.E12), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB355039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355039 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355039 Supplier Novus Biologicals Supplier No. NB100306MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATM Monoclonal specifically detects ATM in Canine (Negative), Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen ATM
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10H11.E12
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated, ataxia telangiectasia mutated (includes complementation groups A, C and D), ATC, ATD, ATDC, ATE, ATM serine/threonine kinase, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1
Host Species Mouse
Immunogen ATM [p Ser1981] Antibody (10H11.E12) was made to a synthetic peptide made to a region surrounding the phosphorylated Serine 1981 of human ATM. [UniProt# Q13315]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Hypoxia, Modulation of DNA Pools, p53 Pathway, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 472
Target Species Canine (Negative), Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.