Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATOX1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB112359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB112359 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB112359 Supplier Novus Biologicals Supplier No. NBP297082UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATOX1 Polyclonal specifically detects ATOX1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATOX1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias ATX1, ATX1 (antioxidant protein 1, yeast) homolog 1, ATX1 antioxidant protein 1 homolog (yeast), copper transport protein ATOX1, hah1, HAH1 MGC138453, Metal transport protein ATX1, MGC138455
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ATOX1 (NP_004036.1; Met1-Glu68).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Golgi Apparatus Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 475
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.