Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP Citrate Lyase Antibody (3A5), Novus Biologicals™
SDP

Catalog No. NB297916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297916 50 μL
NB297917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297916 Supplier Novus Biologicals Supplier No. NBP32653050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ATP Citrate Lyase Monoclonal antibody specifically detects ATP Citrate Lyase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry

Specifications

Antigen ATP Citrate Lyase
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3A5
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias ACLEC 2.3.3.8, ATP citrate lyase, ATP citrate synthase, ATP-citrate (pro-S-)-lyase, ATP-citrate synthase, ATPCL, Citrate cleavage enzyme, CLATP
Host Species Rabbit
Immunogen A synthesized peptide derived from Human ATP Citrate Lyase [UniProt P53396]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 47
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.