Learn More
Description
Specifications
Specifications
| Antigen | ATP10D |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q9P241 |
| Gene Alias | ATPase, class V, type 10D, ATPVDtype IV aminophospholipid transporting ATPase, EC 3.6.3, EC 3.6.3.1, KIAA1487ATPase class V type 10D, probable phospholipid-transporting ATPase VD |
| Gene Symbols | ATP10D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
