Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP10D Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen ATP10D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159982
SDP
View Documents
Novus Biologicals
NBP159982
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

ATP10D Polyclonal specifically detects ATP10D in Human samples. It is validated for Western Blot.
Specifications

Specifications

ATP10D
Polyclonal
Rabbit
Q9P241
57205
Synthetic peptides corresponding to ATP10D(ATPase, class V, type 10D) The peptide sequence was selected from the C terminal of ATP10D. Peptide sequence LFTSAPPVIYGVLEKDVSAETLMQLPELYRSGQKSEAYLPHTFWITLLDA.
Primary
Western Blot
Unconjugated
RUO
ATPase, class V, type 10D, ATPVDtype IV aminophospholipid transporting ATPase, EC 3.6.3, EC 3.6.3.1, KIAA1487ATPase class V type 10D, probable phospholipid-transporting ATPase VD
ATP10D
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.