Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP12A Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB132918 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB132918 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB132918 Supplier Novus Biologicals Supplier No. NBP306153AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP12A Polyclonal specifically detects ATP12A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATP12A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ATP1AL1non-gastric H(+)/K(+) ATPase alpha subunit, ATPase, H+/K+ transporting, nongastric, alpha polypeptide, ATPase, Na+/K+ transporting, alpha polypeptide-like 1, EC 3.6.3, EC 3.6.3.10, Non-gastric H(+)/K(+) ATPase subunit alpha, potassium-transporting ATPase alpha chain 2, Proton pump
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP12A.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 479
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.