Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP13A2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB339339 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339339 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339339 Supplier Novus Biologicals Supplier No. NB11041486MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP13A2 Polyclonal antibody specifically detects ATP13A2 in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunohistochemistry

Specifications

Antigen ATP13A2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Simple Western, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias ATPase type 13A2, EC 3.6.3, EC 3.6.3.-, EC 3.6.3.5, EC 3.6.3.8, FLJ26510, HSA9947, KRPPD, PARK9putative ATPase, Parkinson disease (autosomal recessive) 9 (Kufor-Rakeb syndrome), probable cation-transporting ATPase 13A2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human ATP13A2 protein (within residues 200-300). [Swiss-Prot# Q9NQ11]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neurodegeneration, Neuroscience, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23400
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.