Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    ATP13A2 Antibody, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB11041486
Description
ATP13A2 Polyclonal specifically detects ATP13A2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
| ATP13A2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| ATPase type 13A2, EC 3.6.3, EC 3.6.3.-, EC 3.6.3.5, EC 3.6.3.8, FLJ26510, HSA9947, KRPPD, PARK9putative ATPase, Parkinson disease (autosomal recessive) 9 (Kufor-Rakeb syndrome), probable cation-transporting ATPase 13A2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG | 
| Western Blot, Immunohistochemistry | |
| 0.75 mg/mL | |
| Western Blot 2ug/ ml, Simple Western 1:100, Immunohistochemistry 10 ug/ml | |
| Q9NQ11 | |
| ATP13A2 | |
| A synthetic peptide made to an internal region of the human ATP13A2 protein (within residues 200-300). [Swiss-Prot# Q9NQ11] | |
| 0.1 mL | |
| Cancer, Neurodegeneration, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| 23400 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            For Research Use Only
Spot an opportunity for improvement?Share a Content Correction